(R)-Quinuclidin-3-amine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250225-100MGEA 621 DKK
BLDPBD250225-100MG BLDPBD250225-250MG
(R)-Quinuclidin-3-amine 98%
(R)-Quinuclidin-3-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₁₄N₂
Molekylvægt: 126,19 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08669630
CAS-nummer: 123536-15-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR