(4-Bromobut-1-yn-1-yl)trimethylsilane 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250205-5GEA 1290 DKK
BLDPBD250205-5G BLDPBD250205-250MG BLDPBD250205-1G
(4-Bromobut-1-yn-1-yl)trimethylsilane 95%
(4-Bromobut-1-yn-1-yl)trimethylsilane

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₁₃BrSi
Molekylvægt: 205,16 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD24458737
CAS-nummer: 69361-41-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR