7-Bromo-2-methylbenzo[b]thiophene 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250189-100MGEA 861 DKK
BLDPBD250189-100MG
7-Bromo-2-methylbenzo[b]thiophene 95%
7-Bromo-2-methylbenzo[b]thiophene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₇BrS
Molekylvægt: 227,12 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08706243
CAS-nummer: 75288-49-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR