3-Methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250106-250MGEA 1070 DKK
BLDPBD250106-250MG BLDPBD250106-1G BLDPBD250106-5G BLDPBD250106-100MG
3-Methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%
3-Methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₆H₇N₅
Molekylvægt: 149,153 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD13813768
CAS-nummer: 5399-44-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR