3-Oxo-1-phenyl-2,7,10-trioxa-4-azadodecan-12-oic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250102-5GEA 1680 DKK
BLDPBD250102-5G BLDPBD250102-1G BLDPBD250102-250MG BLDPBD250102-100MG
3-Oxo-1-phenyl-2,7,10-trioxa-4-azadodecan-12-oic acid 95%
3-Oxo-1-phenyl-2,7,10-trioxa-4-azadodecan-12-oic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₁₉NO₆
Molekylvægt: 297,30 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD23379879
CAS-nummer: 165454-06-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR