2-Bromo-5,6-dihydrobenzo[d]thiazol-7(4H)-one 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250043-100MGEA 357 DKK
BLDPBD250043-100MG BLDPBD250043-250MG BLDPBD250043-1G
2-Bromo-5,6-dihydrobenzo[d]thiazol-7(4H)-one 98%
2-Bromo-5,6-dihydrobenzo[d]thiazol-7(4H)-one

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₆BrNOS
Molekylvægt: 232,09 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD13250122
CAS-nummer: 1201633-72-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR