Cresol red sodium salt indicator

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD249917-25GEA 194 DKK
BLDPBD249917-25G BLDPBD249917-500G BLDPBD249917-100G BLDPBD249917-5G
Cresol red sodium salt indicator
Cresol red sodium salt

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₁H₁₇NaO₅S
Molekylvægt: 404,42 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001618
CAS-nummer: 62625-29-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR