8-Acetyl-6-(benzyloxy)-2H-benzo[b][1,4]oxazin-3(4H)-one 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD249764-1GEA 931 DKK
BLDPBD249764-1G BLDPBD249764-250MG BLDPBD249764-5G BLDPBD249764-100MG
8-Acetyl-6-(benzyloxy)-2H-benzo[b][1,4]oxazin-3(4H)-one 97%
8-Acetyl-6-(benzyloxy)-2H-benzo[b][1,4]oxazin-3(4H)-one

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₇H₁₅NO₄
Molekylvægt: 297,305 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD24038851
CAS-nummer: 869478-09-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR