Ethyl 4-((tert-butyldimethylsilyl)oxy)cyclohexanecarboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD249656-1GEA 745 DKK
BLDPBD249656-1G BLDPBD249656-250MG BLDPBD249656-5G BLDPBD249656-100MG
Ethyl 4-((tert-butyldimethylsilyl)oxy)cyclohexanecarboxylate 95%
Ethyl 4-((tert-butyldimethylsilyl)oxy)cyclohexanecarboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₃₀O₃Si
Molekylvægt: 286,48 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD24038913
CAS-nummer: 676560-15-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR