tert-Butyl (2-amino-4-(thiophen-2-yl)phenyl)carbamate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD249510-1GEA 1560 DKK
BLDPBD249510-1G BLDPBD249510-250MG BLDPBD249510-100MG
tert-Butyl (2-amino-4-(thiophen-2-yl)phenyl)carbamate 95%
tert-Butyl (2-amino-4-(thiophen-2-yl)phenyl)carbamate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₅H₁₈N₂O₂S
Molekylvægt: 290,38 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD24038830
CAS-nummer: 335255-43-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR