2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD249400-1GEA 249 DKK
BLDPBD249400-1G BLDPBD249400-250MG BLDPBD249400-5G
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione 98%
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₁₄O₃
Molekylvægt: 182,22 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00467663
CAS-nummer: 1755-15-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR