Methyl 3-(bromomethyl)-4-methoxybenzoate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD249356-1GEA 2240 DKK
BLDPBD249356-1G BLDPBD249356-250MG BLDPBD249356-100MG
Methyl 3-(bromomethyl)-4-methoxybenzoate 98%
Methyl 3-(bromomethyl)-4-methoxybenzoate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₁₁BrO₃
Molekylvægt: 259,1 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD14706047
CAS-nummer: 142031-88-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR