2-(2-(Benzyloxy)-5-(tert-butyl)phenyl)-2-methylpropanenitrile 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD249249-1GEA 1040 DKK
BLDPBD249249-1G BLDPBD249249-100MG BLDPBD249249-250MG
2-(2-(Benzyloxy)-5-(tert-butyl)phenyl)-2-methylpropanenitrile 98%
2-(2-(Benzyloxy)-5-(tert-butyl)phenyl)-2-methylpropanenitrile

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₁H₂₅NO
Molekylvægt: 307,42 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26393887
CAS-nummer: 1246213-27-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR