1-(N-Methoxycarbonyl)indole-2-boronic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD249062-250MGEA 85.3 DKK
BLDPBD249062-250MG BLDPBD249062-1G
1-(N-Methoxycarbonyl)indole-2-boronic acid 95%
1-(N-Methoxycarbonyl)indole-2-boronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₀H₁₀BNO₄
Molekylvægt: 219,01 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD22123264
CAS-nummer: 1001162-89-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR