6-Iodobenzo[d][1,3]dioxol-5-amine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD249061-100MGEA 473 DKK
BLDPBD249061-100MG BLDPBD249061-250MG BLDPBD249061-1G
6-Iodobenzo[d][1,3]dioxol-5-amine 95%
6-Iodobenzo[d][1,3]dioxol-5-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₆INO₂
Molekylvægt: 263,033 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD23728743
CAS-nummer: 1000802-34-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR