Thieno[3,2-c]pyridine-2-carbaldehyde 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD249024-250MGEA 1110 DKK
BLDPBD249024-250MG BLDPBD249024-100MG
Thieno[3,2-c]pyridine-2-carbaldehyde 98%
Thieno[3,2-c]pyridine-2-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₅NOS
Molekylvægt: 163,20 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18453108
CAS-nummer: 94226-19-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR