5-Chloropyridine-3,4-dicarboxylic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD248920-250MGEA 1800 DKK
BLDPBD248920-250MG BLDPBD248920-100MG BLDPBD248920-1G
5-Chloropyridine-3,4-dicarboxylic acid 95%
5-Chloropyridine-3,4-dicarboxylic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₄ClNO₄
Molekylvægt: 201,56 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 101420-62-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR