5-Bromo-4-iodonicotinic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD248909-1GEA 1490 DKK
BLDPBD248909-1G BLDPBD248909-250MG BLDPBD248909-100MG
5-Bromo-4-iodonicotinic acid 95%
5-Bromo-4-iodonicotinic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₃BrINO₂
Molekylvægt: 327,90 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11656277
CAS-nummer: 491588-98-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR