4,5,6,7-Tetrafluoro-1H-benzo[d][1,2,3]triazole 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD248823-250MGEA 752 DKK
BLDPBD248823-250MG BLDPBD248823-5G BLDPBD248823-100MG BLDPBD248823-1G
4,5,6,7-Tetrafluoro-1H-benzo[d][1,2,3]triazole 97%
4,5,6,7-Tetrafluoro-1H-benzo[d][1,2,3]triazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆HF₄N₃
Molekylvægt: 191,09 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01935458
CAS-nummer: 26888-72-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR