Sodium 2-ethylhexyl sulphate 40% in water

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD248481-1000GEA 295 DKK
BLDPBD248481-1000G BLDPBD248481-500G BLDPBD248481-100G
Sodium 2-ethylhexyl sulphate 40% in water
Sodium 2-ethylhexyl sulphate

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₇NaO₄S
Molekylvægt: 232,27 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00042047
CAS-nummer: 126-92-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR