(1S,4S)-2,5-Diphenylbicyclo[2.2.2]octa-2,5-diene 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD248385-100MGEA 1230 DKK
BLDPBD248385-100MG
(1S,4S)-2,5-Diphenylbicyclo[2.2.2]octa-2,5-diene 98%
(1S,4S)-2,5-Diphenylbicyclo[2.2.2]octa-2,5-diene

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₂₀H₁₈
Molekylvægt: 258,357 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD15143645
CAS-nummer: 850409-83-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR