(S)-Methyl 4-(1-aminoethyl)benzoate hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD248303-10GEA 2150 DKK
BLDPBD248303-10G BLDPBD248303-1G BLDPBD248303-0.025 BLDPBD248303-5G BLDPBD248303-250MG BLDPBD248303-25G BLDPBD248303-100MG
(S)-Methyl 4-(1-aminoethyl)benzoate hydrochloride 97%
(S)-Methyl 4-(1-aminoethyl)benzoate hydrochloride

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₄ClNO₂
Molekylvægt: 215,67 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11505983
CAS-nummer: 847728-91-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR