2,3-Dibromotoluene 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD248243-1GEA 621 DKK
BLDPBD248243-1G BLDPBD248243-250MG
2,3-Dibromotoluene 95%
2,3-Dibromotoluene

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₆Br₂
Molekylvægt: 249,9305 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11656704
CAS-nummer: 61563-25-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR