Methyl 3-bromo-5-fluoropyridine-2-carboxylate 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD248140-250MGEA 1070 DKK
BLDPBD248140-250MG BLDPBD248140-5G BLDPBD248140-100MG BLDPBD248140-1G
Methyl 3-bromo-5-fluoropyridine-2-carboxylate 96%
Methyl 3-bromo-5-fluoropyridine-2-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₅BrFNO₂
Molekylvægt: 234,023 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14698117
CAS-nummer: 1214337-00-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR