1-(2,2-Diethoxyethyl)pyrrolidine 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD247984-5GEA 6580 DKK
BLDPBD247984-5G BLDPBD247984-250MG BLDPBD247984-1G BLDPBD247984-100MG
1-(2,2-Diethoxyethyl)pyrrolidine 95+%
1-(2,2-Diethoxyethyl)pyrrolidine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₂₁NO₂
Molekylvægt: 187,27 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 23098-07-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR