3-Chloro-1-methyl-4-nitro-1H-pyrazole 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD247960-5GEA 5150 DKK
BLDPBD247960-5G BLDPBD247960-1G BLDPBD247960-100MG BLDPBD247960-250MG
3-Chloro-1-methyl-4-nitro-1H-pyrazole 95%
3-Chloro-1-methyl-4-nitro-1H-pyrazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₄H₄ClN₃O₂
Molekylvægt: 161,55 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00463968
CAS-nummer: 299930-70-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR