1,2-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD247827-250MGEA 3160 DKK
BLDPBD247827-250MG BLDPBD247827-100MG BLDPBD247827-1G
1,2-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole 98%
1,2-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₁₉BN₂O₂
Molekylvægt: 222,09 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12405495
CAS-nummer: 942070-74-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR