2-(3-Chlorophenyl)thiazole-4-carbaldehyde 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD24777-1GEA 2350 DKK
BLDPBD24777-1G BLDPBD24777-100MG BLDPBD24777-250MG
2-(3-Chlorophenyl)thiazole-4-carbaldehyde 96%
2-(3-Chlorophenyl)thiazole-4-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₆ClNOS
Molekylvægt: 223,68 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06738360
CAS-nummer: 859850-99-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR