3-Chloro-2-methoxy-5-nitropyridine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD247723-5GEA 1560 DKK
BLDPBD247723-5G BLDPBD247723-250MG BLDPBD247723-1G
3-Chloro-2-methoxy-5-nitropyridine 97%
3-Chloro-2-methoxy-5-nitropyridine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₅ClN₂O₃
Molekylvægt: 188,569 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18254096
CAS-nummer: 22353-53-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR