Ethyl 2-(3-bromophenyl)thiazole-4-carboxylate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD24772-5GEA 411 DKK
BLDPBD24772-5G BLDPBD24772-1G
Ethyl 2-(3-bromophenyl)thiazole-4-carboxylate 98%
Ethyl 2-(3-bromophenyl)thiazole-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₀BrNO₂S
Molekylvægt: 312,18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06738338
CAS-nummer: 786654-97-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR