3-Phenylisothiazol-5-amine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD247715-100MGEA 714 DKK
BLDPBD247715-100MG BLDPBD247715-250MG BLDPBD247715-1G
3-Phenylisothiazol-5-amine 95%
3-Phenylisothiazol-5-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₈N₂S
Molekylvægt: 176,24 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07382685
CAS-nummer: 14208-52-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR