5-Bromoisoquinolin-6-amine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD247590-250MGEA 380 DKK
BLDPBD247590-250MG BLDPBD247590-1G BLDPBD247590-100MG BLDPBD247590-5G
5-Bromoisoquinolin-6-amine 98%
5-Bromoisoquinolin-6-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₇BrN₂
Molekylvægt: 223,07 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11847824
CAS-nummer: 566943-98-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR