5-Bromopyridazin-4-amine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD247589-100MGEA 334 DKK
BLDPBD247589-100MG BLDPBD247589-5G BLDPBD247589-250MG BLDPBD247589-25G BLDPBD247589-0.025 BLDPBD247589-1G
5-Bromopyridazin-4-amine 98%
5-Bromopyridazin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₄H₄BrN₃
Molekylvægt: 173,99 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18909369
CAS-nummer: 55928-90-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR