Sodium 3-formylbenzenesulphonate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD247346-100MGEA 621 DKK
BLDPBD247346-100MG BLDPBD247346-250MG BLDPBD247346-1G
Sodium 3-formylbenzenesulphonate 95%
Sodium 3-formylbenzenesulphonate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₅NaO₄S
Molekylvægt: 208,16 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09863878
CAS-nummer: 5330-48-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR