2,4,6-Trichloro-5-methoxypyrimidine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD247148-25GEA 20200 DKK
BLDPBD247148-25G BLDPBD247148-1G BLDPBD247148-250MG BLDPBD247148-100MG BLDPBD247148-5G
2,4,6-Trichloro-5-methoxypyrimidine 95%
2,4,6-Trichloro-5-methoxypyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₅H₃Cl₃N₂O
Molekylvægt: 213,44 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11226168
CAS-nummer: 60703-46-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR