Tetrakis(4-bromophenyl)methane 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD247137-250MGEA 256 DKK
BLDPBD247137-250MG BLDPBD247137-1G BLDPBD247137-100MG BLDPBD247137-25G BLDPBD247137-5G
Tetrakis(4-bromophenyl)methane 95%
Tetrakis(4-bromophenyl)methane

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₅H₁₆Br₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD19688472
CAS-nummer: 105309-59-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR