1-(3-Chlorophenyl)-1H-pyrazole-4-carboxylic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD246936-100MGEA 590 DKK
BLDPBD246936-100MG BLDPBD246936-1G BLDPBD246936-250MG
1-(3-Chlorophenyl)-1H-pyrazole-4-carboxylic acid 95%
1-(3-Chlorophenyl)-1H-pyrazole-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₇ClN₂O₂
Molekylvægt: 222,63 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09951096
CAS-nummer: 1053085-34-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR