2,7-Diazaspiro[4.4]nonan-3-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD246912-250MGEA 1560 DKK
BLDPBD246912-250MG BLDPBD246912-100MG BLDPBD246912-1G
2,7-Diazaspiro[4.4]nonan-3-one 95%
2,7-Diazaspiro[4.4]nonan-3-one
Formel: C₇H₁₂N₂O
Molekylvægt: 140,18 g/mol
MDL Number: MFCD19226811
CAS-nummer: 1226550-00-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR