5-Chloro-1H-pyrazol-3-amine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD246704-1GEA 543 DKK
BLDPBD246704-1G BLDPBD246704-5G BLDPBD246704-250MG BLDPBD246704-10G
5-Chloro-1H-pyrazol-3-amine 98%
5-Chloro-1H-pyrazol-3-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₃H₄ClN₃
Molekylvægt: 117,537 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD26518660
CAS-nummer: 916211-79-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR