2-Formylterephthalic acid 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD246531-250MGEA 497 DKK
BLDPBD246531-250MG BLDPBD246531-1G BLDPBD246531-5G
2-Formylterephthalic acid 95+%
2-Formylterephthalic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₆O₅
Molekylvægt: 194,14 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 69526-90-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR