Ethyl 6-(bromomethyl)picolinate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD246433-100MGEA 721 DKK
BLDPBD246433-100MG BLDPBD246433-1G BLDPBD246433-250MG
Ethyl 6-(bromomethyl)picolinate 98%
Ethyl 6-(bromomethyl)picolinate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₉H₁₀BrNO₂
Molekylvægt: 244,08 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD09026929
CAS-nummer: 97278-44-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR