3,5-Difluoro-4-(trifluoromethoxy)phenol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD246210-100MGEA 1260 DKK
BLDPBD246210-100MG
3,5-Difluoro-4-(trifluoromethoxy)phenol 95%
3,5-Difluoro-4-(trifluoromethoxy)phenol

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₃F₅O₂
Molekylvægt: 214,09 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD23111299
CAS-nummer: 195206-85-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR