5-Cyclopropyl-3-(2,6-dichlorophenyl)isoxazole-4-methanol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD246177-25GEA 1650 DKK
BLDPBD246177-25G BLDPBD246177-1G BLDPBD246177-5G
5-Cyclopropyl-3-(2,6-dichlorophenyl)isoxazole-4-methanol 95%
5-Cyclopropyl-3-(2,6-dichlorophenyl)isoxazole-4-methanol

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₃H₁₁Cl₂NO₂
Molekylvægt: 284,14 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11046189
CAS-nummer: 946426-89-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR