Chroman-7-carboxylic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD246066-100MGEA 1170 DKK
BLDPBD246066-100MG BLDPBD246066-250MG
Chroman-7-carboxylic acid 95%
Chroman-7-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₀O₃
Molekylvægt: 178,18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14706361
CAS-nummer: 527681-33-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR