3-Bromo-6-chloro-2-methylimidazo[1,2-b]pyridazine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245976-100MGEA 504 DKK
BLDPBD245976-100MG
3-Bromo-6-chloro-2-methylimidazo[1,2-b]pyridazine 98%
3-Bromo-6-chloro-2-methylimidazo[1,2-b]pyridazine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₅BrClN₃
Molekylvægt: 246,49 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD21340162
CAS-nummer: 18112-31-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR