Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD24596-5GEA 1490 DKK
BLDPBD24596-5G BLDPBD24596-25G BLDPBD24596-10G BLDPBD24596-250MG BLDPBD24596-1G
Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate 95%
Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₈N₂O₂
Molekylvægt: 176,1700 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08457785
CAS-nummer: 849067-96-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR