3-Bromo-5-fluorobenzene-1,2-diamine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245870-1GEA 225 DKK
BLDPBD245870-1G BLDPBD245870-5G
3-Bromo-5-fluorobenzene-1,2-diamine 97%
3-Bromo-5-fluorobenzene-1,2-diamine
Formel: C₆H₆BrFN₂
Molekylvægt: 205,028 g/mol
MDL Number: MFCD08741398
CAS-nummer: 115440-10-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR