6-Fluoropyridine-2-carboxamide 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245427-250MGEA 140 DKK
BLDPBD245427-250MG BLDPBD245427-1G BLDPBD245427-5G
6-Fluoropyridine-2-carboxamide 98%
6-Fluoropyridine-2-carboxamide

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₅FN₂O
Molekylvægt: 140,115 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04038712
CAS-nummer: 369-03-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR