Methyl 3-Bromo-5-(hydroxymethyl)benzoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245415-100MGEA 396 DKK
BLDPBD245415-100MG BLDPBD245415-1G BLDPBD245415-250MG BLDPBD245415-10G BLDPBD245415-5G
Methyl 3-Bromo-5-(hydroxymethyl)benzoate 95%
Methyl 3-Bromo-5-(hydroxymethyl)benzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉BrO₃
Molekylvægt: 245,07 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11040228
CAS-nummer: 307353-32-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR