2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245280-1GEA 1670 DKK
BLDPBD245280-1G BLDPBD245280-100MG BLDPBD245280-5G BLDPBD245280-250MG BLDPBD245280-10G
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 97%
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₄BF₂NO₂
Molekylvægt: 265,067 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD22419275
CAS-nummer: 1003298-73-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR